Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other KCNMA1 SIRNA Assays ›
          Gene Symbol:
          Kcnma1
          Gene Name:
          potassium large conductance calcium-activated channel, subfamily M, alpha member 1
          Gene Aliases:
          5730414M22Rik, BKCa, MaxiK, Slo, Slo1, mSlo, mSlo1
          Chromosome Location:
          Chr.14: 23292291 - 24004951 on Build GRCm38
          UniGene ID:
          Mm.343607
          Species:
          Mouse
          siRNA ID MSS205723
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Kcnma1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001253358.1 NP_001240287.1 12, 13 1601
          NM_001253359.1 NP_001240288.1 12, 13 1601
          NM_001253360.1 NP_001240289.1 12, 13 1601
          NM_001253361.1 NP_001240290.1 12, 13 1601
          NM_001253362.1 NP_001240291.1 12, 13 1601
          NM_001253363.1 NP_001240292.1 12, 13 1601
          NM_001253364.1 NP_001240293.1 12, 13 1601
          NM_001253365.1 NP_001240294.1 12, 13 1601
          NM_001253366.1 NP_001240295.1 12, 13 1601
          NM_001253367.1 NP_001240296.1 12, 13 1601
          NM_001253368.1 NP_001240297.1 12, 13 1601
          NM_001253369.1 NP_001240298.1 12, 13 1601
          NM_001253370.1 NP_001240299.1 12, 13 1601
          NM_001253371.1 NP_001240300.1 12, 13 1601
          NM_001253372.1 NP_001240301.1 12, 13 1601
          NM_001253373.1 NP_001240302.1 12, 13 1601
          NM_001253374.1 NP_001240303.1 11, 12 1439
          NM_001253375.1 NP_001240304.1 12, 13 1601
          NM_001253376.1 NP_001240305.1 12, 13 1601
          NM_001253377.1 NP_001240306.1 12, 13 1601
          NM_001253378.1 NP_001240307.1 12, 13 1601
          NM_010610.3 NP_034740.2 12, 13 1601
          GenBank mRNA AF156674.1 - 12, 13 1326
          AF465244.1 - 12, 13 1326
          AK142894.1 - 12, 13 1545
          AK149333.1 - 8, 9 1146
          AK166436.1 - 1258
          AK166536.1 - 11, 12 1214
          BC065068.1 - 12, 13 1601
          BC128331.1 - 12, 13 1323
          FJ872117.1 - 12, 13 1326
          HQ221743.1 - 12, 13 1326
          HQ221744.1 - 11, 12 1164
          HQ221745.1 - 12, 13 1326
          HQ221746.1 - 12, 13 1326
          HQ221747.1 - 12, 13 1326
          HQ221748.1 - 12, 13 1326
          HQ221749.1 - 12, 13 1326
          HQ221750.1 - 12, 13 1326
          HQ221751.1 - 12, 13 1326
          HQ221752.1 - 12, 13 1326
          HQ221753.1 - 12, 13 1326
          HQ221754.1 - 12, 13 1326
          HQ221755.1 - 12, 13 1326
          HQ221756.1 - 12, 13 1326
          HQ221757.1 - 12, 13 1326
          HQ221758.1 - 12, 13 1326
          HQ221759.1 - 12, 13 1326
          HQ221760.1 - 12, 13 1326
          HQ221761.1 - 12, 13 1326
          HQ221762.1 - 12, 13 1326
          HQ221763.1 - 12, 13 1326
          HQ529469.1 - 13, 14 1418
          HQ529471.1 - 12, 13 1326
          HQ529475.1 - 12, 13 1326
          HQ529477.1 - 12, 13 1326
          L16912.1 - 12, 13 1407
          U09383.1 - 12, 13 2266

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          transport
          ion transport
          potassium ion transport
          response to osmotic stress
          chemical synaptic transmission
          sensory perception of sound
          circadian rhythm
          adult walking behavior
          response to pH
          neuronal action potential
          cellular potassium ion homeostasis
          response to corticosteroid
          regulation of aldosterone metabolic process
          response to carbon monoxide
          regulation of ion transmembrane transport
          vasodilation
          regulation of vasodilation
          regulation of membrane potential
          auditory receptor cell differentiation
          positive regulation of apoptotic process
          response to estrogen
          locomotor rhythm
          negative regulation of cell volume
          saliva secretion
          cell maturation
          neuromuscular process controlling balance
          protein homooligomerization
          protein homotetramerization
          response to calcium ion
          transmembrane transport
          micturition
          eye blink reflex
          smooth muscle contraction involved in micturition
          relaxation of vascular smooth muscle
          potassium ion transmembrane transport
          actin binding
          ion channel activity
          voltage-gated ion channel activity
          voltage-gated potassium channel activity
          potassium channel activity
          protein binding
          calcium-activated potassium channel activity
          protein complex binding
          protein homodimerization activity
          metal ion binding
          large conductance calcium-activated potassium channel activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-9blpc:80/100.66.131.91:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0