Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Derl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291146.1 | NP_001278075.1 | 5 | 776 | ||
NM_001291147.1 | NP_001278076.1 | 5 | 520 | |||
NM_001291148.1 | NP_001278077.1 | 5 | 482 | |||
NM_033562.4 | NP_291040.1 | 5 | 485 | |||
GenBank mRNA | AF208064.1 | - | 5 | 424 | ||
AK048627.1 | - | 5 | 432 | |||
AK077659.1 | - | 5 | 436 | |||
AK082564.1 | - | 5 | 445 | |||
AK083632.1 | - | 5 | 424 | |||
AK083725.1 | - | 2 | 3188 | |||
AK090105.1 | - | 5 | 494 | |||
AK151934.1 | - | 5 | 776 | |||
BC005682.1 | - | 5 | 424 | 3154448 | ||
BC037503.1 | - | 1 | 67 |