Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Art3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181728.2 | NP_859417.2 | 4 | 302 | ||
GenBank mRNA | AJ311769.1 | - | 2 | 86 | ||
AJ311770.1 | - | 2 | 86 | |||
AJ311771.1 | - | 2 | 86 | |||
AJ311772.1 | - | 2 | 86 | |||
AJ311773.1 | - | 2 | 86 | |||
AK016257.1 | - | 4 | 303 | |||
BC141069.1 | - | 2 | 162 | 9055681 | ||
BC145327.1 | - | 2 | 162 | 9053883 | ||
DQ519368.1 | - | 4 | 301 | |||
DQ519369.1 | - | 4 | 250 | |||
DQ519370.1 | - | 3 | 179 | |||
DQ519371.1 | - | 2 | 378 | |||
DQ810295.1 | - | 3 | 166 |