Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc38a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166456.1 | NP_001159928.1 | 5 | 670 | ||
NM_001166458.1 | NP_001159930.1 | 5 | 877 | |||
NM_134086.4 | NP_598847.2 | 5 | 1363 | |||
GenBank mRNA | AF184240.2 | - | 4 | 455 | ||
AK029189.1 | - | 5 | 540 | |||
AK034130.1 | - | 5 | 1365 | |||
AK035098.1 | - | 4 | 382 | |||
AK047459.1 | - | 5 | 894 | |||
AK049173.1 | - | 5 | 577 | |||
AK049294.1 | - | 5 | 643 | |||
AK050914.1 | - | 5 | 578 | |||
AK081724.1 | - | 5 | 879 | |||
AK082683.1 | - | 5 | 578 | |||
AK162612.1 | - | 5 | 728 | |||
AK165188.1 | - | 5 | 930 | |||
BC030378.1 | - | 5 | 579 | 5375319 |