Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CEP170 SIRNA Assays ›
Gene Symbol:
CEP170
Gene Name:
centrosomal protein 170
Gene Aliases:
FAM68A, KAB, KIAA0470
Chromosome Location:
Chr.1: 243124428 - 243255936 on Build GRCh38
UniGene ID:
Hs.533635
Species:
Human
siRNA ID
HSS190672
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CEP170
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001042404.1
NP_001035863.1
15
4213
NM_001042405.1
NP_001035864.1
15
4135
NM_014812.2
NP_055627.2
16
4507
XM_006711843.3
XP_006711906.1
15
5246
XM_011544334.2
XP_011542636.1
15
5253
XM_011544335.2
XP_011542637.1
15
4666
XM_011544336.2
XP_011542638.1
15
5572
XM_011544337.2
XP_011542639.1
15
5223
XM_011544338.2
XP_011542640.1
15
5172
XM_011544339.2
XP_011542641.1
15
5145
XM_011544340.2
XP_011542642.1
15
5065
XM_011544341.2
XP_011542643.1
16
5017
XM_011544342.2
XP_011542644.1
15
5014
XM_011544343.2
XP_011542645.1
14
4726
XM_011544344.2
XP_011542646.1
15
4773
XM_017002932.1
XP_016858421.1
15
5166
XM_017002933.1
XP_016858422.1
16
4837
XM_017002934.1
XP_016858423.1
16
4480
XM_017002935.1
XP_016858424.1
16
4936
XM_017002936.1
XP_016858425.1
16
4450
XM_017002937.1
XP_016858426.1
16
4909
XM_017002938.1
XP_016858427.1
16
4879
XM_017002939.1
XP_016858428.1
16
4756
XM_017002940.1
XP_016858429.1
16
4399
XM_017002941.1
XP_016858430.1
16
4729
XM_017002942.1
XP_016858431.1
16
4828
XM_017002943.1
XP_016858432.1
16
4648
XM_017002944.1
XP_016858433.1
15
4766
XM_017002945.1
XP_016858434.1
15
4692
XM_017002946.1
XP_016858435.1
15
4540
XM_017002947.1
XP_016858436.1
15
4635
XM_017002948.1
XP_016858437.1
15
4628
XM_017002949.1
XP_016858438.1
15
4584
XM_017002950.1
XP_016858439.1
15
4459
XM_017002951.1
XP_016858440.1
14
4246
GenBank mRNA
AB007939.1
-
14
3780
AB022657.1
-
15
4152
AB022658.1
-
14
3858
AB022659.1
-
14
3780
AK001664.1
-
4
1795
AK313722.1
-
3
297
AL161964.1
-
1
296
BC078155.1
-
4
752
BC140794.1
-
14
3849
9021662
BC146790.1
-
14
3780
9007254
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-858577f6d7-cpd7z:80/100.66.134.75:80.
git-commit: 8a4c7f5d60a2609f21a9257632c46d3db2b553cc
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.34.0-2025.08.32-1.0