Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM214B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317991.1 | NP_001304920.1 | 3 | 663 | ||
NM_025182.3 | NP_079458.2 | 3 | 664 | |||
XM_005251588.1 | XP_005251645.1 | 3 | 798 | |||
XM_005251590.1 | XP_005251647.1 | 3 | 601 | |||
XM_005251591.1 | XP_005251648.1 | 3 | 637 | |||
XM_011518037.1 | XP_011516339.1 | 3 | 620 | |||
XM_011518039.1 | XP_011516341.1 | 3 | 798 | |||
XM_011518040.1 | XP_011516342.1 | 3 | 798 | |||
XM_017015169.1 | XP_016870658.1 | 3 | 741 | |||
GenBank mRNA | AB040972.1 | - | 2 | 3394 | ||
AK021622.1 | - | 3 | 765 | |||
AL583909.1 | - | 3 | 663 | |||
BC004406.1 | - | 3 | 649 | 3638496 |