Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RUFY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040451.2 | NP_001035541.1 | 7 | 1009 | ||
NM_001040452.2 | NP_001035542.1 | 8 | 820 | |||
NM_025158.4 | NP_079434.3 | 8 | 995 | |||
XM_005265993.4 | XP_005266050.1 | 5 | 366 | |||
XM_006714921.3 | XP_006714984.2 | 7 | 939 | |||
XM_006714922.3 | XP_006714985.1 | 4 | 566 | |||
XM_017009891.1 | XP_016865380.1 | 7 | 754 | |||
XM_017009892.1 | XP_016865381.1 | 6 | 792 | |||
XM_017009893.1 | XP_016865382.1 | 5 | 726 | |||
XM_017009894.1 | XP_016865383.1 | 8 | 728 | |||
XM_017009895.1 | XP_016865384.1 | 7 | 800 | |||
GenBank mRNA | AB209507.1 | - | 8 | 720 | ||
AF312367.1 | - | 8 | 986 | |||
AF361055.2 | - | 8 | 968 | |||
AK025904.1 | - | 2 | 58 | |||
AK075021.1 | - | 7 | 909 | |||
AK291926.1 | - | 7 | 1009 | |||
BC032571.1 | - | 8 | 739 | 5243223 |