Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPBAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320728.1 | NP_001307657.1 | 2, 3 | 403 | ||
NM_024610.5 | NP_078886.2 | 2, 3 | 395 | |||
XM_005247762.2 | XP_005247819.1 | 2, 3 | 395 | |||
XM_011513130.2 | XP_011511432.1 | 3, 4 | 313 | |||
GenBank mRNA | AF400663.1 | - | 2, 3 | 362 | ||
AK096705.1 | - | 2, 3 | 290 | |||
AK289410.1 | - | 2, 3 | 395 | |||
BC011897.2 | - | 2, 3 | 379 | 3949150 | ||
BC063629.1 | - | 2, 3 | 335 | |||
BI912690.1 | - | 2, 3 | 375 | |||
HY020808.1 | - | 2, 3 | 406 |