Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF674 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039891.2 | NP_001034980.1 | 6 | 1511 | ||
NM_001146291.1 | NP_001139763.1 | 6 | 1493 | |||
NM_001190417.1 | NP_001177346.1 | 6 | 1496 | |||
XM_011543941.2 | XP_011542243.1 | 6 | 1508 | |||
XM_011543943.2 | XP_011542245.1 | 6 | 1510 | |||
XM_011543944.2 | XP_011542246.1 | 4 | 1318 | |||
XM_011543945.2 | XP_011542247.1 | 3 | 1267 | |||
XM_017029728.1 | XP_016885217.1 | 7 | 1663 | |||
XM_017029729.1 | XP_016885218.1 | 7 | 1685 | |||
XM_017029730.1 | XP_016885219.1 | 7 | 1682 | |||
XM_017029731.1 | XP_016885220.1 | 3 | 1267 | |||
XM_017029732.1 | XP_016885221.1 | 7 | 1660 | |||
XM_017029733.1 | XP_016885222.1 | 7 | 1662 | |||
GenBank mRNA | AK295054.1 | - | 6 | 1425 | ||
AY971607.1 | - | 6 | 1495 | |||
BX648127.1 | - | 1 | 827 |