Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM193B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190946.2 | NP_001177875.1 | 3 | 621 | ||
NR_024019.3 | 4 | 1792 | ||||
XM_006714875.2 | XP_006714938.1 | 3 | 509 | |||
XM_006714876.1 | XP_006714939.1 | 3 | 1232 | |||
XM_006714878.1 | XP_006714941.1 | 4 | 1086 | |||
XM_006714879.3 | XP_006714942.1 | 4 | 329 | |||
XM_006714880.1 | XP_006714943.1 | 3 | 1127 | |||
XM_006714886.2 | XP_006714949.1 | 3 | 509 | |||
XM_011534572.1 | XP_011532874.1 | 3 | 404 | |||
XM_011534573.1 | XP_011532875.1 | 4 | 504 | |||
XM_011534574.1 | XP_011532876.1 | 4 | 949 | |||
XM_011534579.2 | XP_011532881.1 | 3 | 268 | |||
XM_011534580.1 | XP_011532882.1 | 4 | 1293 | |||
XM_011534581.1 | XP_011532883.1 | 3 | 1232 | |||
XM_011534582.1 | XP_011532884.1 | 3 | 1232 | |||
XM_011534583.1 | XP_011532885.1 | 3 | 1232 | |||
GenBank mRNA | AB067518.1 | - | 4 | 1749 | ||
AK096032.1 | - | 2 | 203 | |||
BQ228026.1 | - | 4 | 530 | |||
BX440663.2 | - | 3 | 580 | |||
CN299723.1 | - | 3 | 416 |