Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PHF21A SIRNA Assays ›
          Gene Symbol:
          PHF21A
          Gene Name:
          PHD finger protein 21A
          Gene Aliases:
          BHC80, BM-006
          Chromosome Location:
          Chr.11: 45929319 - 46121434 on Build GRCh38
          UniGene ID:
          Hs.502458
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS181994
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PHF21A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001101802.1 NP_001095272.1 10, 11 1721
          NM_016621.3 NP_057705.3 10, 11 1724
          XM_005252965.4 XP_005253022.1 11, 12 1413
          XM_011520157.2 XP_011518459.1 10, 11 1274
          XM_011520158.2 XP_011518460.1 11, 12 1626
          XM_011520160.2 XP_011518462.1 13, 14 1787
          XM_011520161.2 XP_011518463.1 13, 14 1878
          XM_011520162.2 XP_011518464.1 13, 14 1756
          XM_011520164.2 XP_011518466.1 12, 13 1685
          XM_011520165.2 XP_011518467.1 12, 13 1640
          XM_011520166.2 XP_011518468.1 12, 13 1563
          XM_011520167.2 XP_011518469.1 12, 13 1766
          XM_011520168.2 XP_011518470.1 12, 13 1618
          XM_011520169.2 XP_011518471.1 11, 12 1577
          XM_011520173.2 XP_011518475.1 11, 12 1533
          XM_011520174.2 XP_011518476.1 12, 13 1507
          XM_011520175.2 XP_011518477.1 12, 13 1504
          XM_011520179.2 XP_011518481.1 11, 12 1527
          XM_017017885.1 XP_016873374.1 13, 14 1706
          XM_017017886.1 XP_016873375.1 12, 13 1671
          XM_017017887.1 XP_016873376.1 11, 12 1451
          XM_017017888.1 XP_016873377.1 11, 12 1623
          XM_017017889.1 XP_016873378.1 11, 12 1623
          XM_017017890.1 XP_016873379.1 11, 12 1620
          XM_017017891.1 XP_016873380.1 11, 12 1530
          XM_017017892.1 XP_016873381.1 11, 12 1530
          XM_017017893.1 XP_016873382.1 11, 12 1626
          XM_017017894.1 XP_016873383.1 11, 12 1626
          XM_017017895.1 XP_016873384.1 11, 12 1623
          XM_017017896.1 XP_016873385.1 11, 12 1623
          XM_017017897.1 XP_016873386.1 12, 13 1703
          XM_017017898.1 XP_016873387.1 12, 13 1561
          XM_017017899.1 XP_016873388.1 13, 14 1695
          XM_017017900.1 XP_016873389.1 12, 13 1650
          XM_017017901.1 XP_016873390.1 11, 12 1626
          XM_017017902.1 XP_016873391.1 11, 12 1530
          XM_017017903.1 XP_016873392.1 8, 9 1180
          XM_017017904.1 XP_016873393.1 11, 12 1626
          XM_017017905.1 XP_016873394.1 10, 11 1374
          XM_017017906.1 XP_016873395.1 8, 9 1180
          XM_017017907.1 XP_016873396.1 10, 11 1281
          XM_017017908.1 XP_016873397.1 13, 14 1794
          GenBank mRNA AB051483.1 - 10, 11 1468
          AK021530.1 - 10, 11 1372
          AK023258.1 - 12, 13 1485
          AY090777.1 - 8, 9 1100
          BC015714.2 - 11, 12 1385 3908900
          BX648236.1 - 3, 4 1955

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_001109690, NM_001109691, NM_138755, BC019181, AK129424, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          blood coagulation
          histone deacetylation
          RNA polymerase II regulatory region sequence-specific DNA binding
          chromatin binding
          transcription cofactor activity
          histone deacetylase activity
          protein binding
          zinc ion binding
          histone binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-89bfcd4dd-zgn5p:80/100.66.134.75:80.
          git-commit: f1ed3cb73da2dc18796bd96aca0ec275a318700b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0