Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other APBB2 SIRNA Assays ›
Gene Symbol:
APBB2
Gene Name:
amyloid beta precursor protein binding family B member 2
Gene Aliases:
FE65L, FE65L1
Chromosome Location:
Chr.4: 40810027 - 41216714 on Build GRCh38
UniGene ID:
Hs.479602
Species:
Human
siRNA ID
HSS179349
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: APBB2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001166050.1
NP_001159522.1
6, 7
1473
NM_004307.1
NP_004298.1
6, 7
1476
NM_173075.4
NP_775098.2
6, 7
1473
XM_005248101.3
XP_005248158.1
6, 7
1470
XM_006714005.3
XP_006714068.1
6, 7
1473
XM_006714007.3
XP_006714070.1
6, 7
1471
XM_006714008.3
XP_006714071.1
5, 6
1375
XM_006714009.3
XP_006714072.1
5, 6
1373
XM_006714010.3
XP_006714073.1
6, 7
1470
XM_006714011.3
XP_006714074.1
6, 7
1473
XM_006714012.3
XP_006714075.1
6, 7
1470
XM_011513686.2
XP_011511988.1
7, 8
1495
XM_011513687.2
XP_011511989.1
6, 7
1473
XM_011513688.2
XP_011511990.1
6, 7
1473
XM_017008142.1
XP_016863631.1
6, 7
1394
XM_017008143.1
XP_016863632.1
7, 8
1492
XM_017008144.1
XP_016863633.1
5, 6
1372
XM_017008145.1
XP_016863634.1
5, 6
1375
XM_017008146.1
XP_016863635.1
5, 6
1407
XM_017008147.1
XP_016863636.1
5, 6
1402
XM_017008148.1
XP_016863637.1
4, 5
1306
XM_017008149.1
XP_016863638.1
5, 6
1404
XM_017008150.1
XP_016863639.1
6, 7
1394
XM_017008151.1
XP_016863640.1
5, 6
1375
XM_017008152.1
XP_016863641.1
5, 6
1372
XM_017008153.1
XP_016863642.1
6, 7
1394
XM_017008154.1
XP_016863643.1
5, 6
1375
XM_017008155.1
XP_016863644.1
5, 6
1372
XM_017008156.1
XP_016863645.1
5, 6
1407
XM_017008157.1
XP_016863646.1
4, 5
1306
XM_017008158.1
XP_016863647.1
5, 6
1407
XM_017008159.1
XP_016863648.1
6, 7
1470
GenBank mRNA
AK226179.1
-
6, 7
1390
AK289838.1
-
1
23
AK295974.1
-
1, 2
811
AK304247.1
-
6, 7
1344
BC027946.1
-
6, 7
1313
5203925
CF552608.1
-
1, 2
374
DA603009.1
-
1, 2
449
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
neuron migration
regulation of transcription, DNA-templated
cell cycle arrest
axon guidance
extracellular matrix organization
negative regulation of cell growth
intracellular signal transduction
positive regulation of apoptotic process
negative regulation of apoptotic process
beta-amyloid binding
protein binding
transcription factor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.34.0-2025.08.32-1.0