Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA1324L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142749.2 | NP_001136221.1 | 13, 14 | 1905 | ||
NM_001291990.1 | NP_001278919.1 | 13, 14 | 1663 | |||
NM_001291991.1 | NP_001278920.1 | 12, 13 | 1380 | |||
NM_152748.3 | NP_689961.3 | 12, 13 | 1502 | |||
XM_006715894.3 | XP_006715957.1 | 13, 14 | 1605 | |||
XM_011515918.2 | XP_011514220.1 | 13, 14 | 1756 | |||
XM_011515921.1 | XP_011514223.1 | 12, 13 | 1778 | |||
XM_017011843.1 | XP_016867332.1 | 13, 14 | 1901 | |||
XM_017011844.1 | XP_016867333.1 | 12, 13 | 1495 | |||
GenBank mRNA | AK055902.1 | - | 12, 13 | 1361 | ||
AK296250.1 | - | 12, 13 | 1419 | |||
AK303619.1 | - | 13, 14 | 1665 | |||
BC117309.1 | - | 9, 10 | 1050 | 40125860 |