Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SH2D6 SIRNA Assays ›
          Gene Symbol:
          SH2D6
          Gene Name:
          SH2 domain containing 6
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 85418931 - 85437029 on Build GRCh38
          UniGene ID:
          Hs.209542
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS154746
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SH2D6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_201594.2 NP_963888.2 19, 20 1622
          XM_011532783.1 XP_011531085.1 12, 13 1115
          XM_011532796.2 XP_011531098.1 8, 9 536
          XM_017003834.1 XP_016859323.1 15, 16 4996
          XM_017003835.1 XP_016859324.1 15, 16 4423
          XM_017003836.1 XP_016859325.1 14, 15 4747
          XM_017003837.1 XP_016859326.1 15, 16 4865
          XM_017003838.1 XP_016859327.1 14, 15 4484
          XM_017003839.1 XP_016859328.1 14, 15 2039
          XM_017003840.1 XP_016859329.1 14, 15 5095
          XM_017003842.1 XP_016859331.1 16, 17 4844
          XM_017003843.1 XP_016859332.1 15, 16 4729
          XM_017003844.1 XP_016859333.1 16, 17 4055
          XM_017003845.1 XP_016859334.1 16, 17 4497
          XM_017003846.1 XP_016859335.1 16, 17 4790
          XM_017003847.1 XP_016859336.1 13, 14 5015
          XM_017003848.1 XP_016859337.1 13, 14 5013
          XM_017003849.1 XP_016859338.1 15, 16 4775
          XM_017003850.1 XP_016859339.1 15, 16 4675
          XM_017003851.1 XP_016859340.1 13, 14 5036
          XM_017003852.1 XP_016859341.1 12, 13 700
          XM_017003858.1 XP_016859347.1 19, 20 1622
          XM_017003859.1 XP_016859348.1 12, 13 836
          XM_017003860.1 XP_016859349.1 9, 10 578
          GenBank mRNA AF450483.1 - 3, 4 565
          AK093312.1 - 19, 20 1622
          AY517502.1 - 19, 20 1622
          DB507279.3 - 5, 6 370

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s)

          immune response
          transmembrane receptor protein tyrosine kinase signaling pathway
          intracellular signal transduction

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-956b84784-lf4xt:80/100.66.134.75:80.
          git-commit: 47437712f32dbf53ebfc670c484059c2aca19dcb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0