Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TMEM218 SIRNA Assays ›
Gene Symbol:
TMEM218
Gene Name:
transmembrane protein 218
Gene Aliases:
-
Chromosome Location:
Chr.11: 125094370 - 125111763 on Build GRCh38
UniGene ID:
Hs.731899
Species:
Human
siRNA ID
HSS153561
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TMEM218
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001080546.2
NP_001074015.1
4
598
NM_001258238.1
NP_001245167.1
4
476
NM_001258239.1
NP_001245168.1
4
573
NM_001258240.1
NP_001245169.1
6
636
NM_001258241.1
NP_001245170.1
5
577
NM_001258242.1
NP_001245171.1
4
564
NM_001258243.1
NP_001245172.1
5
667
NM_001258244.1
NP_001245173.1
4
467
NM_001258245.1
NP_001245174.1
6
629
NM_001258246.1
NP_001245175.1
4
549
NM_001258247.1
NP_001245176.1
4
452
NR_047587.1
2
212
NR_047589.1
3
281
NR_047591.1
4
340
XM_006718779.3
XP_006718842.1
6
777
XM_006718780.3
XP_006718843.1
5
674
XM_006718781.3
XP_006718844.1
6
731
XM_006718783.3
XP_006718846.1
4
540
XM_017017322.1
XP_016872811.1
5
564
XM_017017323.1
XP_016872812.1
4
490
XM_017017324.1
XP_016872813.1
4
492
GenBank mRNA
AK091324.1
-
1
3775
AL833215.1
-
1
179
BC132716.1
-
4
435
40146738
BC132718.1
-
4
435
40146740
BC144270.1
-
5
538
9052790
BC144277.1
-
4
469
9052797
BE513148.1
-
4
408
BE974033.1
-
2
57
BI547955.1
-
4
226
BI829527.1
-
4
411
BI856825.1
-
4
342
BM042315.1
-
5
428
BM928068.1
-
508
BQ223298.1
-
6
525
BQ225039.1
-
3
198
BQ420663.1
-
4
443
BQ653243.1
-
4
443
BQ681518.1
-
5
426
BU855096.1
-
4
364
DA899308.1
-
4
436
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.34.0-2025.08.32-1.0