Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TMEM161B SIRNA Assays ›
Gene Symbol:
TMEM161B
Gene Name:
transmembrane protein 161B
Gene Aliases:
FLB3342, PRO1313
Chromosome Location:
Chr.5: 88189419 - 88268917 on Build GRCh38
UniGene ID:
Hs.379972
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS152737
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TMEM161B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001289007.1
NP_001275936.1
10
1227
NM_001289008.1
NP_001275937.1
11
1359
NM_153354.4
NP_699185.1
10
1227
NR_110266.1
11
1327
XM_006714553.1
XP_006714616.1
10
1177
XM_006714554.3
XP_006714617.1
9
1114
XM_006714555.3
XP_006714618.1
8
996
XM_006714556.3
XP_006714619.2
8
957
XM_011543201.2
XP_011541503.1
9
1094
XM_011543202.2
XP_011541504.2
8
1639
XM_011543203.2
XP_011541505.2
6
1444
XM_011543204.2
XP_011541506.2
8
912
XM_017009093.1
XP_016864582.1
10
1177
XM_017009094.1
XP_016864583.1
9
1068
XM_017009095.1
XP_016864584.1
10
1177
XM_017009096.1
XP_016864585.1
9
1098
XM_017009097.1
XP_016864586.1
9
1101
XM_017009098.1
XP_016864587.1
9
1046
XM_017009099.1
XP_016864588.1
8
975
XM_017009100.1
XP_016864589.1
9
1068
XM_017009101.1
XP_016864590.1
9
1030
XM_017009102.1
XP_016864591.1
9
1031
XM_017009103.1
XP_016864592.1
9
1098
XM_017009104.1
XP_016864593.1
9
1161
XM_017009105.1
XP_016864594.1
10
1139
XM_017009106.1
XP_016864595.1
9
1031
XM_017009107.1
XP_016864596.1
9
1061
XM_017009108.1
XP_016864597.1
8
946
XM_017009109.1
XP_016864598.1
9
1111
XM_017009110.1
XP_016864599.1
9
1097
GenBank mRNA
AK300006.1
-
6
575
AK300893.1
-
11
1309
AK301091.1
-
10
1178
BC037287.2
-
10
1196
4837154
BX647345.1
-
11
1247
BX648757.1
-
8
5000
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-89bfcd4dd-zgn5p:80/100.66.134.75:80.
git-commit: f1ed3cb73da2dc18796bd96aca0ec275a318700b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.33.0-2025.07.30-1.0