Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NBPF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256416.2 | NP_001243345.1 | 15 | 2125 | ||
NM_001256417.2 | NP_001243346.1 | 14 | 1951 | |||
NM_032264.4 | NP_115640.1 | 15 | 2161 | |||
NR_046176.2 | 18 | 2690 | ||||
XM_006710957.1 | XP_006711020.1 | 15 | 2019 | |||
XM_006710958.1 | XP_006711021.1 | 18 | 2475 | |||
XM_006710959.2 | XP_006711022.1 | 16 | 2259 | |||
XM_011542276.1 | XP_011540578.1 | 19 | 2732 | |||
XM_011542277.1 | XP_011540579.1 | 19 | 2717 | |||
XM_011542278.1 | XP_011540580.1 | 19 | 2546 | |||
XM_011542281.1 | XP_011540583.1 | 16 | 2082 | |||
XM_017002498.1 | XP_016857987.1 | 19 | 2531 | |||
XM_017002499.1 | XP_016857988.1 | 14 | 1797 | |||
XM_017002500.1 | XP_016857989.1 | 18 | 2550 | |||
XM_017002501.1 | XP_016857990.1 | 19 | 2435 | |||
XM_017002502.1 | XP_016857991.1 | 18 | 2364 | |||
GenBank mRNA | AK095602.1 | - | 18 | 2644 | ||
AK292580.1 | - | 15 | 2026 | |||
AK314597.1 | - | 15 | 2089 | |||
AY894564.1 | - | 14 | 1667 |