Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPB7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014424.4 | NP_055239.1 | 2 | 1133 | ||
XM_011541249.1 | XP_011539551.1 | 2 | 1154 | |||
XM_011541250.2 | XP_011539552.1 | 2 | 1151 | |||
GenBank mRNA | AF155908.1 | - | 2 | 380 | ||
AJ243191.1 | - | 2 | 380 | |||
AK057295.1 | - | 2 | 1131 | |||
AK075151.1 | - | 1 | 944 | |||
AK092275.1 | - | 3 | 656 | |||
AK095911.1 | - | 2 | 386 | |||
AL832181.1 | - | 1 | 889 | |||
BC006319.2 | - | 2 | 799 | 4131506 | ||
CR749298.1 | - | 2 | 398 | |||
CR749419.1 | - | 2 | 398 |