Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF226 SIRNA Assays ›
Gene Symbol:
ZNF226
Gene Name:
zinc finger protein 226
Gene Aliases:
-
Chromosome Location:
Chr.19: 44165062 - 44177685 on Build GRCh38
UniGene ID:
Hs.145956
Species:
Human
siRNA ID
HSS144563
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF226
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001032372.1
NP_001027544.1
5
368
NM_001032373.1
NP_001027545.1
5
332
NM_001032374.1
NP_001027546.1
5
501
NM_001146220.2
NP_001139692.1
6
392
NM_001319088.1
NP_001306017.1
5
535
NM_001319089.1
NP_001306018.1
6
392
NM_001319090.1
NP_001306019.1
5
412
NM_015919.3
NP_057003.2
5
332
NM_016444.2
NP_057528.2
6
555
XM_005259227.2
XP_005259284.1
6
666
XM_006723367.3
XP_006723430.1
7
562
XM_006723368.3
XP_006723431.1
7
448
XM_006723369.2
XP_006723432.1
6
505
XM_017027262.1
XP_016882751.1
6
514
XM_017027263.1
XP_016882752.1
5
336
XM_017027264.1
XP_016882753.1
5
493
XM_017027265.1
XP_016882754.1
5
400
GenBank mRNA
AF070666.1
-
5
327
AF228418.1
-
6
553
AK023091.1
-
5
328
AK314724.1
-
6
351
BC017786.1
-
5
324
BC024197.2
-
5
368
3532273
BI457780.1
-
5
333
BX091242.1
-
5
494
BX537487.1
-
5
324
BX648775.1
-
5
496
CD700800.1
-
5
358
DA080156.1
-
5
496
DA178589.1
-
5
355
DA435005.1
-
6
338
DA558473.1
-
5
340
DB015503.1
-
5
521
DR002748.1
-
5
374
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.34.0-2025.08.32-1.0