Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF146 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099638.1 | NP_001093108.1 | 3 | 1865 | ||
NM_001099639.1 | NP_001093109.1 | 3 | 1824 | |||
NM_007145.2 | NP_009076.2 | 4 | 1939 | |||
XM_005259214.2 | XP_005259271.1 | 4 | 1764 | |||
XM_017027244.1 | XP_016882733.1 | 3 | 1390 | |||
XM_017027245.1 | XP_016882734.1 | 4 | 1395 | |||
XM_017027246.1 | XP_016882735.1 | 3 | 1294 | |||
XM_017027247.1 | XP_016882736.1 | 3 | 1356 | |||
GenBank mRNA | AK311486.1 | - | 1 | 409 | ||
AK313460.1 | - | 1 | 877 | |||
BC065568.1 | - | 4 | 1408 | |||
BC110500.1 | - | 1 | 1165 | 40028224 | ||
BC110501.1 | - | 1 | 1165 | 40028226 | ||
X70394.1 | - | 2 | 1272 |