Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ZNF438 SIRNA Assays ›
          Gene Symbol:
          ZNF438
          Gene Name:
          zinc finger protein 438
          Gene Aliases:
          bA330O11.1
          Chromosome Location:
          Chr.10: 30844634 - 31032406 on Build GRCh38
          UniGene ID:
          Hs.660642
          Species:
          Human
          siRNA ID HSS137418
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price 492.00
          Your Price
          Online offer:
          492.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ZNF438
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001143766.1 NP_001137238.1 7 841
          NM_001143767.1 NP_001137239.1 5 654
          NM_001143768.1 NP_001137240.1 6 712
          NM_001143769.1 NP_001137241.1 7 966
          NM_001143770.1 NP_001137242.1 8 892
          NM_001143771.1 NP_001137243.1 7 863
          NM_182755.2 NP_877432.1 7 793
          XM_006717393.3 XP_006717456.1 5 608
          XM_006717396.3 XP_006717459.1 4 542
          XM_006717398.3 XP_006717461.1 8 833
          XM_006717399.3 XP_006717462.1 7 766
          XM_011519371.2 XP_011517673.1 6 2816
          XM_011519372.2 XP_011517674.1 6 843
          XM_011519373.2 XP_011517675.1 5 2758
          XM_011519374.2 XP_011517676.1 3 585
          XM_011519376.2 XP_011517678.1 8 918
          XM_011519377.2 XP_011517679.1 8 914
          XM_017015863.1 XP_016871352.1 3 408
          XM_017015864.1 XP_016871353.1 8 3052
          XM_017015865.1 XP_016871354.1 7 2993
          XM_017015866.1 XP_016871355.1 7 883
          XM_017015867.1 XP_016871356.1 6 2798
          XM_017015868.1 XP_016871357.1 5 1104
          XM_017015869.1 XP_016871358.1 5 2739
          XM_017015870.1 XP_016871359.1 6 1638
          XM_017015871.1 XP_016871360.1 5 1098
          XM_017015872.1 XP_016871361.1 6 2796
          XM_017015873.1 XP_016871362.1 7 2973
          XM_017015874.1 XP_016871363.1 5 659
          GenBank mRNA AF428258.1 - 6 688
          AK131357.1 - 7 966
          AK292730.1 - 7 759
          AL833056.1 - 2 402
          BC101622.1 - 3 418 8069128
          BC104757.1 - 3 418 8143760
          BC143462.1 - 4 517
          BC143464.1 - 4 597
          DQ356011.1 - 7 787
          DQ356012.1 - 7 778

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          DNA binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7967b4b575-m9zbg:80/100.66.134.75:80.
          git-commit: a47d3bef29014c2544d0032e3b149846d456d521
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0