Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FER1L6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039112.2 | NP_001034201.2 | 24 | 3271 | ||
XM_006716618.3 | XP_006716681.1 | 24 | 3198 | |||
XM_011517231.2 | XP_011515533.1 | 25 | 3237 | |||
XM_011517232.2 | XP_011515534.1 | 25 | 3378 | |||
XM_011517233.2 | XP_011515535.1 | 24 | 3339 | |||
XM_011517235.2 | XP_011515537.1 | 25 | 3238 | |||
XM_011517236.2 | XP_011515538.1 | 5 | 966 | |||
GenBank mRNA | AB196633.1 | - | 24 | 3271 |
Gene Symbol: FER1L6-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103547.1 | 2 | 257 | |||
GenBank mRNA | AK057332.1 | - | 2 | 257 | ||
BC101502.1 | - | 2 | 198 | |||
BC101506.1 | - | 2 | 198 |