Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHST9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256316.1 | NP_001243245.1 | 2 | 376 | ||
NM_031422.5 | NP_113610.2 | 2 | 376 | |||
XM_006722555.3 | XP_006722618.1 | 2 | 333 | |||
XM_017026033.1 | XP_016881522.1 | 1 | 275 | |||
XM_017026034.1 | XP_016881523.1 | 2 | 402 | |||
GenBank mRNA | AF239821.1 | - | 2 | 253 | ||
AF332472.1 | - | 1 | 167 | |||
AJ707539.1 | - | 2 | 363 | |||
AK093349.1 | - | 1 | 275 | |||
AY358488.1 | - | 2 | 355 | |||
BC025764.1 | - | 2 | 320 | 5205829 | ||
BI710997.1 | - | 2 | 381 | |||
BM919514.1 | - | 2 | 355 | |||
DB444451.1 | - | 1 | 306 | |||
KU178792.1 | - | 1 | 62 | |||
KU178793.1 | - | 1 | 62 |