Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THAP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008695.1 | NP_001008695.1 | 3, 4 | 269 | ||
NM_030573.2 | NP_085050.2 | 2, 3 | 443 | |||
GenBank mRNA | AK315457.1 | - | 2, 3 | 428 | ||
BC004346.1 | - | 2, 3 | 414 | 3633743 | ||
BE264280.1 | - | 2, 3 | 342 | |||
BI223500.1 | - | 2, 3 | 386 | |||
BM790105.1 | - | 3, 4 | 269 | |||
CR456343.1 | - | 2, 3 | 261 |