Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CAPRIN2 SIRNA Assays ›
Gene Symbol:
CAPRIN2
Gene Name:
caprin family member 2
Gene Aliases:
C1QDC1, EEG-1, EEG1, RNG140
Chromosome Location:
Chr.12: 30709552 - 30754994 on Build GRCh38
UniGene ID:
Hs.234355
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS128054
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CAPRIN2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001002259.2
NP_001002259.1
18
3788
NM_001206856.2
NP_001193785.1
17
3664
NM_001319842.1
NP_001306771.1
17
3702
NM_001319843.1
NP_001306772.1
18
3785
NM_001319844.1
NP_001306773.1
18
2938
NM_001319845.1
NP_001306774.1
19
3043
NM_001319846.1
NP_001306775.1
19
3033
NM_023925.4
NP_076414.2
17
3638
NM_032156.4
NP_115532.3
18
3829
XM_006719138.3
XP_006719201.1
18
3848
XM_006719139.3
XP_006719202.1
17
3806
XM_006719140.3
XP_006719203.1
17
3803
XM_006719142.3
XP_006719205.1
17
3701
XM_006719144.3
XP_006719207.1
16
3659
XM_006719145.3
XP_006719208.1
18
3955
XM_006719146.3
XP_006719209.1
18
3987
XM_006719147.3
XP_006719210.1
18
3892
XM_017019851.1
XP_016875340.1
17
3743
XM_017019852.1
XP_016875341.1
19
3003
XM_017019853.1
XP_016875342.1
19
3073
XM_017019854.1
XP_016875343.1
16
3656
XM_017019855.1
XP_016875344.1
19
2970
XM_017019856.1
XP_016875345.1
18
2928
XM_017019857.1
XP_016875346.1
18
2898
XM_017019858.1
XP_016875347.1
18
2925
XM_017019859.1
XP_016875348.1
17
2754
XM_017019860.1
XP_016875349.1
19
3955
XM_017019861.1
XP_016875350.1
17
2778
XM_017019862.1
XP_016875351.1
19
3952
XM_017019863.1
XP_016875352.1
19
3999
XM_017019865.1
XP_016875354.1
17
3850
XM_017019866.1
XP_016875355.1
19
3050
XM_017019867.1
XP_016875356.1
19
3077
XM_017019868.1
XP_016875357.1
19
3047
XM_017019869.1
XP_016875358.1
19
3074
XM_017019870.1
XP_016875359.1
19
2987
XM_017019871.1
XP_016875360.1
19
3058
XM_017019872.1
XP_016875361.1
18
2909
XM_017019873.1
XP_016875362.1
17
2825
XM_017019874.1
XP_016875363.1
18
2920
XM_017019875.1
XP_016875364.1
17
2762
XM_017019876.1
XP_016875365.1
16
2471
XM_017019877.1
XP_016875366.1
15
2429
XM_017019878.1
XP_016875367.1
15
2426
XM_017019879.1
XP_016875368.1
17
2755
XM_017019880.1
XP_016875369.1
14
2219
XM_017019881.1
XP_016875370.1
18
2964
XM_017019882.1
XP_016875371.1
18
2904
XM_017019883.1
XP_016875372.1
15
2473
GenBank mRNA
AB058776.1
-
3275
AF525416.1
-
10
1299
AK024247.1
-
5
2755
AK026222.1
-
4
604
AL834212.1
-
16
2316
AY074490.1
-
17
3638
AY074491.1
-
17
3702
BC007520.1
-
4
509
BC066295.1
-
4
540
BC111007.1
-
4
592
BX537569.1
-
15
5844
BX538080.1
-
5
724
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of translation
negative regulation of cell growth
positive regulation of protein binding
positive regulation of peptidyl-serine phosphorylation
positive regulation of transcription from RNA polymerase II promoter
positive regulation of dendrite morphogenesis
positive regulation of dendritic spine morphogenesis
positive regulation of canonical Wnt signaling pathway
RNA binding
receptor binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c48b94556-6l4lq:80/100.66.134.75:80.
git-commit: 256cec658627a60c8e1196038bf5905a13cd45ba
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.33.0-2025.07.30-1.0