Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF415 SIRNA Assays ›
Gene Symbol:
ZNF415
Gene Name:
zinc finger protein 415
Gene Aliases:
ZfLp
Chromosome Location:
Chr.19: 53107879 - 53133077 on Build GRCh38
UniGene ID:
Hs.147765
Species:
Human
siRNA ID
HSS125037
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF415
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001136038.2
NP_001129510.2
4
1200
NM_001164309.1
NP_001157781.1
4
1073
NM_018355.3
NP_060825.2
4
968
NR_028343.1
7
1329
XM_006723267.1
XP_006723330.1
6
1286
XM_006723268.1
XP_006723331.1
4
1034
XM_006723272.2
XP_006723335.1
5
1066
XM_006723273.2
XP_006723336.1
5
1150
XM_006723274.2
XP_006723337.1
5
1227
XM_006723277.2
XP_006723340.1
5
1161
XM_006723279.1
XP_006723342.1
5
1320
XM_006723280.2
XP_006723343.1
2
776
XM_006723281.2
XP_006723344.1
3
802
XM_006723282.1
XP_006723345.1
3
1111
XM_011527100.1
XP_011525402.1
5
1310
XM_011527101.2
XP_011525403.1
5
1285
XM_011527103.1
XP_011525405.1
5
1105
XM_011527104.1
XP_011525406.1
5
1399
XM_017026952.1
XP_016882441.1
5
1113
XM_017026953.1
XP_016882442.1
6
1433
XM_017026954.1
XP_016882443.1
6
1310
XM_017026955.1
XP_016882444.1
6
1261
XM_017026956.1
XP_016882445.1
7
1411
XM_017026957.1
XP_016882446.1
6
1149
XM_017026958.1
XP_016882447.1
7
1465
XM_017026959.1
XP_016882448.1
6
1336
XM_017026960.1
XP_016882449.1
5
1051
XM_017026961.1
XP_016882450.1
6
1497
XM_017026962.1
XP_016882451.1
6
1342
XM_017026963.1
XP_016882452.1
5
1238
XM_017026964.1
XP_016882453.1
5
1188
XM_017026965.1
XP_016882454.1
4
1202
XM_017026966.1
XP_016882455.1
5
1330
XM_017026967.1
XP_016882456.1
4
1128
XM_017026968.1
XP_016882457.1
6
1426
XM_017026969.1
XP_016882458.1
2
847
XM_017026970.1
XP_016882459.1
6
1303
XM_017026971.1
XP_016882460.1
6
1522
XM_017026972.1
XP_016882461.1
4
1104
XM_017026973.1
XP_016882462.1
5
1296
XM_017026974.1
XP_016882463.1
6
1375
GenBank mRNA
AK002053.1
-
4
940
AK095504.1
-
7
1301
AY283600.1
-
4
940
BC063880.1
-
5
1151
30388597
BC069668.1
-
5
1150
7262632
DQ925695.1
-
3
802
DQ925696.1
-
5
1139
DQ925697.1
-
6
1149
DQ925698.1
-
5
1251
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.34.0-2025.08.32-1.0