Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF821 SIRNA Assays ›
Gene Symbol:
ZNF821
Gene Name:
zinc finger protein 821
Gene Aliases:
-
Chromosome Location:
Chr.16: 71859680 - 71886224 on Build GRCh38
UniGene ID:
Hs.744055
Species:
Human
siRNA ID
HSS124586
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF821
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001201552.1
NP_001188481.1
6
822
NM_001201553.1
NP_001188482.1
5
807
NM_001201554.1
NP_001188483.1
5
696
NM_001201556.1
NP_001188485.1
6
822
NM_001318238.1
NP_001305167.1
6
776
NM_001318239.1
NP_001305168.1
5
549
NM_017530.2
NP_060000.1
4
681
XM_005256033.2
XP_005256090.1
7
896
XM_006721233.1
XP_006721296.1
6
755
XM_011523211.2
XP_011521513.1
5
601
XM_011523212.2
XP_011521514.1
5
654
XM_011523214.2
XP_011521516.1
5
624
XM_011523215.2
XP_011521517.1
3
295
XM_011523220.2
XP_011521522.1
5
544
XM_017023409.1
XP_016878898.1
6
740
XM_017023410.1
XP_016878899.1
6
1487
XM_017023411.1
XP_016878900.1
6
564
XM_017023412.1
XP_016878901.1
7
896
XM_017023413.1
XP_016878902.1
5
624
XM_017023414.1
XP_016878903.1
5
683
XM_017023415.1
XP_016878904.1
5
621
XM_017023416.1
XP_016878905.1
6
833
XM_017023417.1
XP_016878906.1
6
827
XM_017023418.1
XP_016878907.1
6
689
XM_017023419.1
XP_016878908.1
6
671
XM_017023420.1
XP_016878909.1
7
916
XM_017023421.1
XP_016878910.1
5
682
XM_017023422.1
XP_016878911.1
4
337
XM_017023423.1
XP_016878912.1
6
830
XM_017023424.1
XP_016878913.1
5
684
XM_017023425.1
XP_016878914.1
4
538
GenBank mRNA
AF070588.1
-
4
649
AK131336.1
-
1
323
AK296603.1
-
5
807
AK313693.1
-
5
656
AL538838.3
-
6
841
BC012116.1
-
5
653
4553333
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
transcription factor activity, sequence-specific DNA binding
protein binding
transcription regulatory region DNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.34.0-2025.08.32-1.0