Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PHF21A SIRNA Assays ›
Gene Symbol:
PHF21A
Gene Name:
PHD finger protein 21A
Gene Aliases:
BHC80, BM-006
Chromosome Location:
Chr.11: 45929319 - 46121434 on Build GRCh38
UniGene ID:
Hs.502458
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS122032
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PHF21A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001101802.1
NP_001095272.1
9
1492
NM_016621.3
NP_057705.3
9
1495
XM_005252965.4
XP_005253022.1
10
1184
XM_011520157.2
XP_011518459.1
9
1045
XM_011520158.2
XP_011518460.1
10
1397
XM_011520160.2
XP_011518462.1
12
1558
XM_011520161.2
XP_011518463.1
12
1649
XM_011520162.2
XP_011518464.1
12
1527
XM_011520164.2
XP_011518466.1
11
1456
XM_011520165.2
XP_011518467.1
11
1411
XM_011520166.2
XP_011518468.1
11
1334
XM_011520167.2
XP_011518469.1
11
1537
XM_011520168.2
XP_011518470.1
11
1389
XM_011520169.2
XP_011518471.1
10
1348
XM_011520173.2
XP_011518475.1
10
1304
XM_011520174.2
XP_011518476.1
11
1278
XM_011520175.2
XP_011518477.1
11
1275
XM_011520179.2
XP_011518481.1
10
1298
XM_017017885.1
XP_016873374.1
12
1477
XM_017017886.1
XP_016873375.1
11
1442
XM_017017887.1
XP_016873376.1
10
1222
XM_017017888.1
XP_016873377.1
10
1394
XM_017017889.1
XP_016873378.1
10
1394
XM_017017890.1
XP_016873379.1
10
1391
XM_017017891.1
XP_016873380.1
10
1301
XM_017017892.1
XP_016873381.1
10
1301
XM_017017893.1
XP_016873382.1
10
1397
XM_017017894.1
XP_016873383.1
10
1397
XM_017017895.1
XP_016873384.1
10
1394
XM_017017896.1
XP_016873385.1
10
1394
XM_017017897.1
XP_016873386.1
11
1474
XM_017017898.1
XP_016873387.1
11
1332
XM_017017899.1
XP_016873388.1
12
1466
XM_017017900.1
XP_016873389.1
11
1421
XM_017017901.1
XP_016873390.1
10
1397
XM_017017902.1
XP_016873391.1
10
1301
XM_017017903.1
XP_016873392.1
7
951
XM_017017904.1
XP_016873393.1
10
1397
XM_017017905.1
XP_016873394.1
9
1145
XM_017017906.1
XP_016873395.1
7
951
XM_017017907.1
XP_016873396.1
9
1052
XM_017017908.1
XP_016873397.1
12
1565
GenBank mRNA
AB051483.1
-
9
1239
AF208848.1
-
1
19
AK021530.1
-
9
1143
AK023258.1
-
11
1256
AY090777.1
-
7
871
BC015714.2
-
10
1156
3908900
BX648236.1
-
2
1726
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
blood coagulation
histone deacetylation
RNA polymerase II regulatory region sequence-specific DNA binding
chromatin binding
transcription cofactor activity
histone deacetylase activity
protein binding
zinc ion binding
histone binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-89bfcd4dd-zgn5p:80/100.66.134.75:80.
git-commit: f1ed3cb73da2dc18796bd96aca0ec275a318700b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.33.0-2025.07.30-1.0