Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282454.1 | NP_001269383.1 | 9 | 1046 | ||
NM_014426.3 | NP_055241.1 | 9 | 1174 | |||
NM_152227.2 | NP_689413.1 | 10 | 1129 | |||
GenBank mRNA | AF121855.1 | - | 10 | 985 | ||
AK001793.1 | - | 9 | 891 | |||
AK026227.1 | - | 2 | 86 | |||
AK054634.1 | - | 10 | 1917 | |||
AK129505.1 | - | 9 | 891 | |||
AK223308.1 | - | 9 | 859 | |||
AK296947.1 | - | 10 | 998 | |||
AK308017.1 | - | 10 | 1130 | |||
BC062638.1 | - | 9 | 831 | |||
BC093623.1 | - | 9 | 1100 | 7939468 | ||
BC093980.1 | - | 9 | 1100 | 7939825 | ||
BC143274.1 | - | 9 | 1100 | 9051776 |