Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other COBL SIRNA Assays ›
Gene Symbol:
COBL
Gene Name:
cordon-bleu WH2 repeat protein
Gene Aliases:
-
Chromosome Location:
Chr.7: 51016212 - 51316818 on Build GRCh38
UniGene ID:
Hs.99141
Species:
Human
siRNA ID
HSS118432
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: COBL
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001287436.1
NP_001274365.1
7
1154
NM_001287438.1
NP_001274367.1
7
1154
NM_015198.3
NP_056013.2
7
1154
XM_005271750.1
XP_005271807.1
8
1190
XM_005271751.1
XP_005271808.1
7
1145
XM_005271756.1
XP_005271813.1
7
1145
XM_011515234.1
XP_011513536.1
9
1265
XM_011515235.1
XP_011513537.1
8
1220
XM_011515236.1
XP_011513538.1
9
1265
XM_011515237.1
XP_011513539.1
9
1265
XM_011515238.1
XP_011513540.1
9
1265
XM_011515239.1
XP_011513541.1
9
1265
XM_011515240.1
XP_011513542.1
8
1220
XM_011515241.1
XP_011513543.1
3
269
XM_017011898.1
XP_016867387.1
8
1190
XM_017011899.1
XP_016867388.1
7
1145
XM_017011900.1
XP_016867389.1
7
1145
XM_017011901.1
XP_016867390.1
7
1145
GenBank mRNA
AB014533.1
-
7
1135
AK309254.1
-
7
1145
BC029275.1
-
2
28
BC045771.1
-
5
453
BC094695.1
-
7
1100
BC111496.1
-
7
1154
BC136441.1
-
7
1069
9020430
BC144099.1
-
7
1069
9052619
BC150263.1
-
7
1135
9007207
CR936689.1
-
6
645
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
embryonic axis specification
somite specification
neural tube closure
liver development
actin filament polymerization
notochord development
floor plate development
digestive tract development
collateral sprouting in absence of injury
actin filament network formation
positive regulation of dendrite development
positive regulation of ruffle assembly
actin monomer binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.34.0-2025.08.32-1.0