Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF510 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001314059.1 | NP_001300988.1 | 2 | 678 | ||
NM_001314060.1 | NP_001300989.1 | 2 | 289 | |||
NM_014930.2 | NP_055745.1 | 2 | 289 | |||
XM_005251808.2 | XP_005251865.1 | 2 | 279 | |||
XM_011518393.2 | XP_011516695.1 | 2 | 709 | |||
XM_017014483.1 | XP_016869972.1 | 3 | 341 | |||
GenBank mRNA | AB023189.1 | - | 1 | 206 | ||
AK292600.1 | - | 2 | 259 | |||
BC036676.1 | - | 2 | 676 | 4792684 | ||
BC058022.1 | - | 2 | 253 | |||
BC068587.1 | - | 2 | 672 | 4374918 | ||
DA496439.1 | - | 2 | 227 |