Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168412.1 | NP_001161884.1 | 3, 4 | 518 | ||
NM_005724.5 | NP_005715.1 | 4, 5 | 710 | |||
NM_198902.2 | NP_944492.1 | 3, 4 | 635 | |||
XM_017021857.1 | XP_016877346.1 | 4, 5 | 319 | |||
GenBank mRNA | AF054840.1 | - | 4, 5 | 436 | ||
AF133423.1 | - | 4, 5 | 510 | |||
AK001305.1 | - | 4, 5 | 556 | |||
AK001310.1 | - | 4, 5 | 542 | |||
AK001326.1 | - | 4, 5 | 653 | |||
AK027793.1 | - | 1, 2 | 1761 | |||
AK075336.1 | - | 4, 5 | 591 | |||
AK075482.1 | - | 3, 4 | 516 | |||
AK293677.1 | - | 5, 6 | 499 | |||
AK298157.1 | - | 3, 4 | 399 | |||
BC000704.2 | - | 4, 5 | 611 | 3349181 | ||
BC004280.1 | - | 4, 5 | 572 | 3616380 | ||
BC009248.2 | - | 4, 5 | 575 | 2964568 | ||
BC011206.1 | - | 4, 5 | 604 | 3457158 |