Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF133 SIRNA Assays ›
Gene Symbol:
ZNF133
Gene Name:
zinc finger protein 133
Gene Aliases:
ZNF150, pHZ-13, pHZ-66
Chromosome Location:
Chr.20: 18288228 - 18316996 on Build GRCh38
UniGene ID:
Hs.472221
Species:
Human
siRNA ID
HSS111581
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF133
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001083330.2
NP_001076799.1
4
1180
NM_001282995.1
NP_001269924.1
6
1416
NM_001282996.1
NP_001269925.1
6
1290
NM_001282997.1
NP_001269926.1
5
1341
NM_001282998.1
NP_001269927.1
4
1170
NM_001282999.1
NP_001269928.1
4
1183
NM_001283000.1
NP_001269929.1
5
1359
NM_001283001.1
NP_001269930.1
5
1346
NM_001283002.1
NP_001269931.1
5
1248
NM_001283003.1
NP_001269932.1
5
1321
NM_001283004.1
NP_001269933.1
5
1308
NM_001283005.1
NP_001269934.1
2
944
NM_001283006.1
NP_001269935.1
3
1022
NM_001283007.1
NP_001269936.1
3
1040
NM_001283008.1
NP_001269937.1
4
1210
NM_003434.5
NP_003425.2
7
1592
XM_005260819.1
XP_005260876.1
4
992
XM_005260820.4
XP_005260877.1
6
1249
XM_011529336.1
XP_011527638.1
7
1328
XM_011529337.1
XP_011527639.1
6
1200
XM_011529338.2
XP_011527640.1
6
1194
XM_011529339.1
XP_011527641.1
7
1328
XM_011529340.1
XP_011527642.1
2
803
XM_017028041.1
XP_016883530.1
5
9093
XM_017028042.1
XP_016883531.1
5
2367
XM_017028043.1
XP_016883532.1
5
9090
XM_017028044.1
XP_016883533.1
7
9440
XM_017028045.1
XP_016883534.1
5
9508
XM_017028046.1
XP_016883535.1
5
9106
XM_017028047.1
XP_016883536.1
5
2460
XM_017028048.1
XP_016883537.1
4
2289
XM_017028049.1
XP_016883538.1
7
9450
XM_017028050.1
XP_016883539.1
7
1348
XM_017028051.1
XP_016883540.1
8
1524
XM_017028052.1
XP_016883541.1
6
1188
XM_017028053.1
XP_016883542.1
4
1222
XM_017028054.1
XP_016883543.1
5
1102
XM_017028055.1
XP_016883544.1
8
1518
XM_017028056.1
XP_016883545.1
10
1737
XM_017028057.1
XP_016883546.1
7
1305
XM_017028058.1
XP_016883547.1
8
1522
GenBank mRNA
AK123005.1
-
2
2758
AK291389.1
-
5
1098
AK294263.1
-
3
771
AK295273.1
-
5
1020
AK295513.1
-
5
1084
AK313525.1
-
4
919
AK316386.1
-
2
698
BC001887.1
-
7
1398
3535533
BT007310.1
-
3
839
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.34.0-2025.08.32-1.0