Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CTAGE5 SIRNA Assays ›
Gene Symbol:
CTAGE5
Gene Name:
CTAGE family member 5, ER export factor
Gene Aliases:
MEA6, MGEA, MGEA11, MGEA6
Chromosome Location:
Chr.14: 39265264 - 39351193 on Build GRCh38
UniGene ID:
Hs.509200
Species:
Human
siRNA ID
HSS106514
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
492.00
Your Price
Online offer:
492.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CTAGE5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001247988.1
NP_001234917.1
19, 20
1873
NM_001247989.1
NP_001234918.1
20, 21
2186
NM_001247990.1
NP_001234919.1
20, 21
2076
NM_005930.3
NP_005921.2
20, 21
2171
NM_203354.2
NP_976229.1
20, 21
1922
NM_203355.2
NP_976230.1
19, 20
2042
NM_203356.2
NP_976231.1
20, 21
2002
XM_005267648.4
XP_005267705.1
20, 21
1882
XM_006720148.2
XP_006720211.1
20, 21
1884
XM_011536775.2
XP_011535077.1
21, 22
2249
XM_011536778.2
XP_011535080.1
21, 22
1937
XM_011536784.2
XP_011535086.1
20, 21
1800
XM_011536785.2
XP_011535087.1
20, 21
1792
XM_017021314.1
XP_016876803.1
21, 22
2265
XM_017021315.1
XP_016876804.1
21, 22
1966
XM_017021316.1
XP_016876805.1
21, 22
1953
XM_017021317.1
XP_016876806.1
21, 22
2374
XM_017021318.1
XP_016876807.1
21, 22
1995
XM_017021319.1
XP_016876808.1
21, 22
1931
XM_017021320.1
XP_016876809.1
20, 21
1957
XM_017021321.1
XP_016876810.1
20, 21
2121
XM_017021322.1
XP_016876811.1
19, 20
1801
XM_017021323.1
XP_016876812.1
21, 22
1749
XM_017021324.1
XP_016876813.1
21, 22
1837
XM_017021325.1
XP_016876814.1
19, 20
1761
XM_017021326.1
XP_016876815.1
19, 20
1671
XM_017021327.1
XP_016876816.1
19, 20
1663
XM_017021328.1
XP_016876817.1
19, 20
1664
XM_017021329.1
XP_016876818.1
19, 20
1643
XM_017021330.1
XP_016876819.1
21, 22
1874
GenBank mRNA
AB209528.1
-
19, 20
2480
AF338233.1
-
20, 21
2016
AF338234.1
-
20, 21
2049
AK091252.1
-
19, 20
2251
AK298935.1
-
19, 20
1825
AK315254.1
-
19, 20
1763
AL832233.1
-
19, 20
1863
AL833415.1
-
18, 19
1624
BC039017.1
-
20, 21
1964
BC051363.1
-
20, 21
1789
4576721
BC064355.1
-
20, 21
2132
4803802
BX640994.1
-
20, 21
1904
BX648014.1
-
20, 21
1904
U73682.1
-
19, 20
1752
U94780.1
-
20, 21
2149
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
ER to Golgi vesicle-mediated transport
protein exit from endoplasmic reticulum
cargo loading into vesicle
positive regulation of catalytic activity
protein localization to endoplasmic reticulum exit site
regulation of guanyl-nucleotide exchange factor activity
protein binding
enzyme activator activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7967b4b575-m9zbg:80/100.66.134.75:80.
git-commit: a47d3bef29014c2544d0032e3b149846d456d521
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.34.0-2025.08.32-1.0