Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh2d4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177816.3 | NP_808484.2 | 7 | 1107 | ||
GenBank mRNA | AK050671.1 | - | 4 | 821 | ||
AK134033.1 | - | 4 | 597 | |||
AX776199.1 | - | 7 | 1107 | |||
BC138964.1 | - | 7 | 1205 | 8861986 |