Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDHA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000284.3 | NP_000275.1 | 3 | 326 | ||
NM_001173454.1 | NP_001166925.1 | 4 | 440 | |||
NM_001173455.1 | NP_001166926.1 | 3 | 326 | |||
NM_001173456.1 | NP_001166927.1 | 3 | 326 | |||
XM_017029574.1 | XP_016885063.1 | 4 | 427 | |||
GenBank mRNA | AK129887.1 | - | 3 | 261 | ||
AK222740.1 | - | 3 | 257 | |||
AK222988.1 | - | 3 | 291 | |||
AK293250.1 | - | 4 | 405 | |||
AK296341.1 | - | 3 | 292 | |||
AK296457.1 | - | 3 | 240 | |||
AK312263.1 | - | 3 | 292 | |||
BC002406.1 | - | 3 | 238 | 2961286 | ||
DC361191.1 | - | 3 | 326 | |||
EF590117.1 | - | 358 | ||||
J03503.1 | - | 3 | 253 | |||
J03575.1 | - | 3 | 226 | |||
L13318.1 | - | 3 | 286 | |||
L48690.1 | - | 3 | 286 | |||
M24848.1 | - | 3 | 224 | |||
X52709.1 | - | 3 | 235 | |||
X52710.1 | - | 3 | 237 |