Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026585.3 | NP_080861.2 | 3 | 244 | ||
GenBank mRNA | AK011156.1 | - | 3 | 190 | ||
AK038318.1 | - | 3 | 222 | |||
AK122320.1 | - | 2 | 250 | |||
AK145716.1 | - | 3 | 244 | |||
AK161181.1 | - | 3 | 193 | |||
AK163765.1 | - | 2 | 232 | |||
AK168388.1 | - | 3 | 231 | |||
AK215654.1 | - | 1 | 31 | |||
BC032177.1 | - | 1 | 471 | |||
BC040393.1 | - | 3 | 169 | |||
BC049979.1 | - | 3 | 220 | |||
BC056942.1 | - | 3 | 171 | 6827177 |