Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pqlc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164420.1 | NP_001157892.1 | 5 | 1061 | ||
NM_001164421.1 | NP_001157893.1 | 5 | 993 | |||
NM_001164422.1 | NP_001157894.1 | 5 | 988 | |||
NM_025861.3 | NP_080137.2 | 6 | 1047 | |||
GenBank mRNA | AK009256.1 | - | 5 | 983 | ||
AK018759.1 | - | 5 | 1425 | |||
AK153023.1 | - | 5 | 1013 | |||
AK153176.1 | - | 5 | 1014 | |||
BC008231.1 | - | 5 | 930 | 2647794 | ||
BC029179.1 | - | 5 | 935 | 3487991 | ||
BC043686.1 | - | 1225 | 5135928 |