Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CREM SIRNA Assays ›
          Gene Symbol:
          Crem
          Gene Name:
          cAMP responsive element modulator
          Gene Aliases:
          Icer
          Chromosome Location:
          Chr.17: 57021704 - 57090888 on Build Rnor_6.0
          UniGene ID:
          Rn.10251
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 59658

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Crem
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001110860.2 NP_001104330.1 9 1168
          NM_001271101.1 NP_001258030.1 9 1168
          NM_001271102.1 NP_001258031.1 2 122
          NM_001271245.1 NP_001258174.1 3 204
          NM_001271246.1 NP_001258175.1 3 266
          NM_001271247.1 NP_001258176.1 3 312
          NM_001271248.1 NP_001258177.1 2 230
          NM_017334.2 NP_059030.2 3 158
          XM_006254049.2 XP_006254111.1 8 1126
          XM_006254050.2 XP_006254112.1 8 1126
          XM_006254051.2 XP_006254113.1 8 952
          XM_006254052.2 XP_006254114.1 7 1284
          XM_006254053.2 XP_006254115.1 8 906
          XM_006254054.2 XP_006254116.1 7 1006
          XM_006254055.2 XP_006254117.1 7 870
          XM_006254056.2 XP_006254118.1 7 870
          XM_006254057.2 XP_006254119.1 8 973
          XM_006254058.2 XP_006254120.1 7 937
          XM_006254059.2 XP_006254121.1 7 937
          XM_006254060.2 XP_006254122.1 6 1137
          XM_006254061.2 XP_006254123.1 6 1137
          XM_006254062.2 XP_006254124.1 5 1101
          XM_006254063.2 XP_006254125.1 6 856
          XM_006254064.2 XP_006254126.1 7 717
          XM_006254065.2 XP_006254127.1 7 717
          XM_006254066.2 XP_006254128.1 6 681
          XM_006254067.2 XP_006254129.1 6 681
          XM_006254068.2 XP_006254130.1 4 912
          XM_006254069.2 XP_006254131.1 3 545
          XM_006254070.2 XP_006254132.1 2 1030
          XM_008771691.1 XP_008769913.1 3 591
          XM_008771692.1 XP_008769914.1 3 187
          XM_008771693.1 XP_008769915.1 2 829
          XM_008771694.1 XP_008769916.1 2 166
          XM_008771695.1 XP_008769917.1 2 145
          GenBank mRNA AB031423.1 - 3 187
          BC078899.1 - 9 1168 7133070
          CO556486.1 - 2 179
          FM033648.1 - 3 302
          FM089481.1 - 3 267
          FQ213544.1 - 2 122
          FQ219343.1 - 2 119
          S66024.1 - 3 155
          U04835.1 - 3 196
          Z15158.1 - 7 761

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_001110857, NM_001271506, NM_001110858, NM_001271503, NM_001110854, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          glucose metabolic process
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          fatty acid metabolic process
          glycosphingolipid metabolic process
          spermatogenesis
          cAMP-mediated signaling
          circadian regulation of gene expression
          regulation of circadian rhythm
          negative regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          retinoic acid receptor signaling pathway
          response to cAMP
          core promoter sequence-specific DNA binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          sequence-specific DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-775c674fdc-h55f5:80/100.66.134.75:80.
          git-commit: 140417879117b66a3780ee196c9dcc5b0ff3ca88
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0