Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Enpp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057104.2 | NP_476445.2 | 4 | 438 | ||
GenBank mRNA | BC081747.1 | - | 4 | 398 | 7115236 | |
D28560.1 | - | 4 | 438 | |||
DQ131564.1 | - | 4 | 438 |