Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vamp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057097.2 | NP_476438.1 | 3 | 176 | ||
GenBank mRNA | BC088119.1 | - | 3 | 176 | 7366421 | |
S63830.1 | - | 228 |