Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019253.3 | NP_062126.3 | 4, 5 | 1059 | ||
XM_006229252.2 | XP_006229314.1 | 9, 10 | 1964 | |||
XM_006229253.2 | XP_006229315.1 | 9, 10 | 1905 | |||
XM_006229254.2 | XP_006229316.1 | 8, 9 | 1804 | |||
XM_006229255.2 | XP_006229317.1 | 8, 9 | 1880 | |||
XM_006229257.2 | XP_006229319.1 | 7, 8 | 1711 | |||
XM_008759346.1 | XP_008757568.1 | 9, 10 | 1439 | |||
GenBank mRNA | BC092196.1 | - | 2, 3 | 424 | 7313460 | |
FQ211673.1 | - | 9, 10 | 1568 | |||
FQ213308.1 | - | 6, 7 | 873 | |||
S49400.1 | - | 4, 5 | 1059 |