Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700017N19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081246.1 | NP_001074715.1 | 3, 4 | 292 | ||
GenBank mRNA | AK006074.1 | - | 2, 3 | 263 | ||
AK014917.1 | - | 3, 4 | 270 |