Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A730020M07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK042746.1 | - | 4 | 1872 | ||
BE647989.1 | - | 1 | 240 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | NM_176992.1 | - | 1871 | Chr. - - [-] on Build |