Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173118.2 | NP_775141.2 | 4 | 506 | ||
GenBank mRNA | AJ515237.1 | - | 4 | 441 | ||
BC058132.1 | - | 4 | 469 | 6920130 | ||
FQ214040.1 | - | 4 | 427 | |||
FQ216545.1 | - | 4 | 431 | |||
FQ219301.1 | - | 4 | 448 | |||
FQ219863.1 | - | 4 | 439 | |||
FQ220660.1 | - | 4 | 427 | |||
FQ220919.1 | - | 4 | 433 | |||
FQ228456.1 | - | 4 | 431 | |||
FQ229422.1 | - | 4 | 486 | |||
FQ229782.1 | - | 4 | 427 | |||
FQ229783.1 | - | 4 | 429 | |||
FQ230107.1 | - | 4 | 433 |