Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF220 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319956.1 | NP_001306885.1 | 15 | 2547 | ||
NM_001319957.1 | NP_001306886.1 | 14 | 1832 | |||
NM_018150.3 | NP_060620.2 | 15 | 2551 | |||
XM_005270992.1 | XP_005271049.1 | 15 | 2620 | |||
XM_005270993.1 | XP_005271050.1 | 15 | 2538 | |||
XM_005270996.1 | XP_005271053.1 | 11 | 1487 | |||
XM_011541698.1 | XP_011540000.1 | 15 | 2314 | |||
XM_011541699.1 | XP_011540001.1 | 15 | 2578 | |||
XM_011541700.1 | XP_011540002.1 | 15 | 2357 | |||
XM_011541702.1 | XP_011540004.1 | 14 | 1749 | |||
XM_017001623.1 | XP_016857112.1 | 15 | 2500 | |||
XM_017001624.1 | XP_016857113.1 | 15 | 2514 | |||
XM_017001625.1 | XP_016857114.1 | 15 | 2258 | |||
XM_017001626.1 | XP_016857115.1 | 13 | 1563 | |||
GenBank mRNA | AF151080.1 | - | 1 | 176 | ||
AK001459.1 | - | 9 | 1908 | |||
AK025744.1 | - | 14 | 1640 | |||
AK056424.1 | - | 15 | 2458 | |||
AK124790.1 | - | 6 | 3249 | |||
AK225180.1 | - | 11 | 1182 | |||
BC000279.1 | - | 1 | 61 | |||
BC034221.1 | - | 15 | 2423 | 4814781 | ||
BC073835.1 | - | 4 | 649 | |||
BC098266.1 | - | 15 | 2221 | 40026694 | ||
BC098300.1 | - | 15 | 2221 | 40026695 | ||
BC105790.1 | - | 1 | 76 |