Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SYTL2 SIRNA Assays ›
Gene Symbol:
SYTL2
Gene Name:
synaptotagmin like 2
Gene Aliases:
CHR11SYT, EXO4, PPP1R151, SGA72M, SLP2, SLP2A
Chromosome Location:
Chr.11: 85694221 - 85811182 on Build GRCh38
UniGene ID:
Hs.369520
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
43795
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SYTL2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001162951.2
NP_001156423.1
14
2590
NM_001162952.2
NP_001156424.1
6
1120
NM_001162953.2
NP_001156425.1
14
2593
NM_001289608.1
NP_001276537.1
15
2399
NM_001289609.1
NP_001276538.1
7
1233
NM_001289610.1
NP_001276539.1
7
804
NM_032943.4
NP_116561.1
15
2769
NM_206929.3
NP_996812.1
8
924
NM_206930.3
NP_996813.1
5
1097
XM_005274057.1
XP_005274114.1
14
6229
XM_005274060.1
XP_005274117.1
15
2413
XM_005274061.1
XP_005274118.1
15
2410
XM_005274062.2
XP_005274119.1
16
2834
XM_005274063.1
XP_005274120.1
14
2293
XM_005274066.1
XP_005274123.1
13
2242
XM_005274067.4
XP_005274124.1
7
2536
XM_011545105.1
XP_011543407.1
15
6277
XM_011545106.1
XP_011543408.1
15
6274
XM_011545107.2
XP_011543409.1
16
6698
XM_011545108.1
XP_011543410.1
14
6157
XM_011545109.2
XP_011543411.1
15
6259
XM_011545114.2
XP_011543416.1
6
2442
XM_011545115.1
XP_011543417.1
4
536
XM_011545116.2
XP_011543418.1
5
2321
XM_017017933.1
XP_016873422.1
14
6226
XM_017017934.1
XP_016873423.1
16
6695
XM_017017935.1
XP_016873424.1
12
6073
XM_017017936.1
XP_016873425.1
12
5771
XM_017017937.1
XP_016873426.1
15
2314
XM_017017938.1
XP_016873427.1
14
2272
XM_017017939.1
XP_016873428.1
8
2580
XM_017017940.1
XP_016873429.1
7
2532
GenBank mRNA
AB046817.1
-
15
2780
AK024872.1
-
5
1261
AK056722.1
-
1
1882
AK074737.1
-
8
881
AK092121.1
-
5
915
AK124754.1
-
8
2505
AK127504.1
-
9
3467
AK131365.1
-
8
1397
AK222707.1
-
5
1261
AK222710.1
-
5
1261
AK296224.1
-
15
2381
AK296740.1
-
12
2329
AK298604.1
-
7
722
AL834422.1
-
2
159
AY386362.1
-
8
4784
BC015540.2
-
5
591
BC068495.1
-
7
958
BC110315.1
-
6
649
BC136450.1
-
14
2525
9020439
BC144114.1
-
14
2525
9052634
BC144115.1
-
14
2522
9052635
BX647671.1
-
9
3220
BX648068.1
-
8
2666
EU035829.1
-
14
2246
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
intracellular protein transport
exocytosis
vesicle docking involved in exocytosis
vesicle fusion
negative regulation of phosphatase activity
vesicle-mediated transport
regulation of calcium ion-dependent exocytosis
calcium ion-regulated exocytosis of neurotransmitter
positive regulation of mucus secretion
protein localization to plasma membrane
phosphatidylserine binding
calcium ion binding
protein binding
calcium-dependent phospholipid binding
phosphatidylinositol-4,5-bisphosphate binding
Rab GTPase binding
phosphatase binding
syntaxin binding
clathrin binding
neurexin family protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.34.0-2025.08.32-1.0