Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TMEM161B SIRNA Assays ›
          Gene Symbol:
          TMEM161B
          Gene Name:
          transmembrane protein 161B
          Gene Aliases:
          FLB3342, PRO1313
          Chromosome Location:
          Chr.5: 88189419 - 88268917 on Build GRCh38
          UniGene ID:
          Hs.379972
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 40145

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TMEM161B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001289007.1 NP_001275936.1 3 321
          NM_001289008.1 NP_001275937.1 4 453
          NM_153354.4 NP_699185.1 3 321
          NR_110266.1 3 321
          XM_006714553.1 XP_006714616.1 3 271
          XM_006714554.3 XP_006714617.1 2 208
          XM_006714555.3 XP_006714618.1 2 188
          XM_011543201.2 XP_011541503.1 2 188
          XM_011543202.2 XP_011541504.2 2 831
          XM_011543204.2 XP_011541506.2 2 142
          XM_017009093.1 XP_016864582.1 3 271
          XM_017009094.1 XP_016864583.1 2 162
          XM_017009095.1 XP_016864584.1 3 271
          XM_017009096.1 XP_016864585.1 3 290
          XM_017009097.1 XP_016864586.1 2 195
          XM_017009098.1 XP_016864587.1 2 140
          XM_017009099.1 XP_016864588.1 2 167
          XM_017009100.1 XP_016864589.1 2 162
          XM_017009101.1 XP_016864590.1 2 162
          XM_017009102.1 XP_016864591.1 3 261
          XM_017009103.1 XP_016864592.1 3 290
          XM_017009104.1 XP_016864593.1 2 195
          XM_017009105.1 XP_016864594.1 3 271
          XM_017009106.1 XP_016864595.1 3 261
          XM_017009107.1 XP_016864596.1 2 193
          XM_017009108.1 XP_016864597.1 2 176
          XM_017009109.1 XP_016864598.1 3 303
          XM_017009110.1 XP_016864599.1 2 191
          GenBank mRNA AK299896.1 - 3 261
          AK300893.1 - 4 403
          AK301091.1 - 3 272
          AY358693.1 - 3 279
          BC037287.2 - 3 290 4837154
          BX647345.1 - 3 241
          BX648757.1 - 2 221

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-89bfcd4dd-zgn5p:80/100.66.134.75:80.
          git-commit: f1ed3cb73da2dc18796bd96aca0ec275a318700b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0