Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF182 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165032.1 | NP_001158504.1 | 4 | 709 | ||
NM_001165033.1 | NP_001158505.1 | 2 | 488 | |||
NM_001165034.1 | NP_001158506.1 | 3 | 596 | |||
NM_152737.3 | NP_689950.1 | 3 | 626 | |||
XM_017010448.1 | XP_016865937.1 | 4 | 646 | |||
XM_017010449.1 | XP_016865938.1 | 3 | 710 | |||
XM_017010450.1 | XP_016865939.1 | 2 | 694 | |||
GenBank mRNA | AK090576.1 | - | 2 | 445 | ||
AK315528.1 | - | 3 | 607 | |||
BC030666.1 | - | 3 | 526 | 5259219 | ||
BC050030.1 | - | 2 | 350 | 5297801 | ||
DB176198.1 | - | 2 | 488 |