Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SMTN SIRNA Assays ›
Gene Symbol:
SMTN
Gene Name:
smoothelin
Gene Aliases:
-
Chromosome Location:
Chr.22: 31081296 - 31104624 on Build GRCh38
UniGene ID:
Hs.149098
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
36636
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SMTN
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001207017.1
NP_001193946.1
4, 5
510
NM_001207018.1
NP_001193947.1
3, 4
532
NM_006932.4
NP_008863.3
4, 5
528
NM_134269.2
NP_599031.1
4, 5
528
NM_134270.2
NP_599032.2
4, 5
528
XM_006724298.1
XP_006724361.1
4, 5
506
XM_006724302.1
XP_006724365.1
4, 5
565
XM_011530333.2
XP_011528635.1
5, 6
611
XM_011530336.2
XP_011528638.1
5, 6
612
XM_011530337.2
XP_011528639.1
5, 6
612
XM_011530338.2
XP_011528640.1
5, 6
611
XM_011530339.2
XP_011528641.1
5, 6
612
XM_011530340.2
XP_011528642.1
5, 6
612
XM_017028913.1
XP_016884402.1
5, 6
612
XM_017028914.1
XP_016884403.1
5, 6
611
XM_017028915.1
XP_016884404.1
5, 6
611
XM_017028916.1
XP_016884405.1
5, 6
612
XM_017028917.1
XP_016884406.1
5, 6
612
XM_017028918.1
XP_016884407.1
5, 6
612
XM_017028919.1
XP_016884408.1
5, 6
611
GenBank mRNA
AB209643.1
-
1, 2
101
AF064238.3
-
4, 5
505
AJ010306.2
-
4, 5
505
AK096953.1
-
4, 5
563
AK128627.1
-
3, 4
2073
AK128719.1
-
4, 5
374
AK295418.1
-
3, 4
532
AK303424.1
-
3, 4
307
AK304084.1
-
4, 5
510
AY061971.1
-
4, 5
506
AY061972.1
-
4, 5
506
BC034237.2
-
4, 5
496
3912887
DC342566.1
-
4, 5
528
Y13492.2
-
4, 5
506
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
smooth muscle contraction
muscle organ development
actin binding
structural constituent of muscle
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-89bfcd4dd-zgn5p:80/100.66.134.75:80.
git-commit: f1ed3cb73da2dc18796bd96aca0ec275a318700b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.33.0-2025.07.30-1.0